Learn More
Medchemexpress LLC Polyvinyl alcohol | 9002-89-5 | 98.0-99.0% | 31,000-50,000 | (C2H4O)x | 1g
SDP
Catalog No. 5000265912
Encompass_Preferred
Polyvinyl alcohol (Mw 31000-50000 98-99% hydrolyzed) also known as PVA is a vinyl water-soluble polymer that can be used as a non-ionic surfactant It can also be used as a biodegradable polymer and can be used in adhesives coatings textiles ceramics and cosmetics Polyvinyl alcohol can be used in tissue engineering by electrospinning Polyvinyl alcohol can achieve high cellular density infiltration and uniform distribution facilitating functional connections between cells Polyvinyl alcohol can improve cell vitality through in vitro cultivation Polyvinyl alcohol demonstrates promising inhibition of ostersarcoma cancer cells with Doxorubicin (HY-15142A)[1][2]
Catalog No. 50-002-65912 Supplier Medchemexpress LLC Supplier No. HYY0850K1G
Polyvinyl alcohol (Mw 31000-50000 98-99% hydrolyzed) also known as PVA is a vinyl water-soluble polymer that can be used as a non-ionic surfactant It can also be used as a biodegradable polymer and can be used in adhesives coatings textiles ceramics and cosmetics Polyvinyl alcohol can be used in tissue engineering by electrospinning Polyvinyl alcohol can achieve high cellular density infiltration and uniform distribution facilitating functional connections between cells Polyvinyl alcohol can improve cell vitality through in vitro cultivation Polyvinyl alcohol demonstrates promising inhibition of ostersarcoma cancer cells with Doxorubicin (HY-15142A)[1][2]