Learn More
Medchemexpress LLC Polyvinyl alcohol | 9002-89-5 | MFCD00081922 | 205000 (Average) | (C2H4O)n | 25g
SDP
Catalog No. 5000268697
Encompass_Preferred
PVA (Polyvinyl alcohol) (Mw 205000) is a polyvinyl alcohol with a molecular weight of 205000 which is used as a biological excipient PVA (Mw 205000) has excellent optical properties high dielectric strength and excellent charge storage and can be used as an embedded pad and substrate for metal/inorganic nanofillers for sensing optoelectronic devices and many other applications[1]
Catalog No. 50-002-68697 Supplier Medchemexpress LLC Supplier No. HYY0850U625G
PVA (Polyvinyl alcohol) (Mw 205000) is a polyvinyl alcohol with a molecular weight of 205000 which is used as a biological excipient PVA (Mw 205000) has excellent optical properties high dielectric strength and excellent charge storage and can be used as an embedded pad and substrate for metal/inorganic nanofillers for sensing optoelectronic devices and many other applications[1]