Learn More
Hach Company Polyvinyl Alcohol Dispersing Agent, 50 mL SCDB
Supplier:  Hach Company 2376526
Encompass
For determination of Ammonia and Total Kjeldahl Nitrogen by the Nessler Method. 50 mL Self-Contained Dropping Bottle.
Catalog No. 50-199-5894