Learn More
Polyvinyl Alcohol, Partially Hydrolyzed, 87.0 to 89.0%, BAKER™, J.T. Baker™
Shop All Avantor ProductsSpecifications
Specifications
| Quantity | 1 kg |
| Flash Point | 79.00°C |
| Packaging | Wide Mouth Amber Glass Bottle |
| Formula Weight | 11,000 to 31,000 |
| Percent Purity | 87.0 to 89.0% |
| Chemical Name or Material | Polyvinyl Alcohol, Partially Hydrolyzed |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.