Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polyvinyl Alcohol, USP, Spectrum™ Chemical
SDP

Catalog No. 18605408 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
50 g
250 g
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-605-408 250 g Poly Bottle
18-605-412 50 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-605-408 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P1180250GM
Only null left
Add to Cart
Add to Cart

P1180, 9002-89-5, (C2H4O)n

Spectrum™ Chemical Polyvinyl Alcohol, USP is used to relieve dry, irritated eyes. All Spectrum Chemical USP are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 9002-89-5
Molecular Formula (C2H4O)n
Molecular Weight (g/mol) 44.05
MDL Number MFCD00081922
InChI Key IMROMDMJAWUWLK-UHFFFAOYSA-N
IUPAC Name ethenol
SMILES OC(-*)C-*

Specifications

pH 5.0 to 8.0
Acid Value 3
CAS Max % 1
Quantity 250 g
Grade USP
Ignition Residue 0.01
Loss on Drying 0.05
Packaging Poly Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.