Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polyvinyl Butyral, Sigma-Aldrich

Catalog No. 01258899 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-899 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-258-899 Supplier MilliporeSigma Supelco Supplier No. P110010100G
Add to Cart
Add to Cart

Polyvinyl butyral resin similar to Butvar™ B-98 for thin film support in electron microscopy.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63148-65-2

Specifications

Physical Form Powder
Quantity 100 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.