Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Polyvinyl chloride | 9002-86-2 | MFCD00084459 | 20,000-80,000 | (C2H3Cl)x | 100 G
SDP

Catalog No. 5000284049
Encompass_Preferred

Chloroethene (polymer), also known as polyvinyl chloride (PVC), is supplied as a research-grade solid powder for analytical and laboratory applications. The product is accompanied by COA, SDS, and handling documentation and is intended for research use only.

  • Research-grade polyvinyl chloride powder for laboratory and analytical use.
  • Molecular formula (C2H3Cl)x with molecular weight range 20,000-80,000.
  • CAS number 9002-86-2 for unambiguous chemical identification.
  • Packaged as 100 G solid powder suitable for small-scale research studies.
  • Provided with COA, datasheet, SDS, and handling instructions to support quality control.
  • Storage: powder stable at -20°C (3 years) or 4°C (2 years).
  • Safety notes: reported cytotoxic and reproductive toxicity; handle per SDS and use appropriate precautions.

Catalog No. 50-002-84049 Supplier Medchemexpress LLC Supplier No. HYW687022A100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chloroethene (polymer), also known as polyvinyl chloride (PVC), is supplied as a research-grade solid powder for analytical and laboratory applications. The product is accompanied by COA, SDS, and handling documentation and is intended for research use only.

  • Research-grade polyvinyl chloride powder for laboratory and analytical use.
  • Molecular formula (C2H3Cl)x with molecular weight range 20,000-80,000.
  • CAS number 9002-86-2 for unambiguous chemical identification.
  • Packaged as 100 G solid powder suitable for small-scale research studies.
  • Provided with COA, datasheet, SDS, and handling instructions to support quality control.
  • Storage: powder stable at -20°C (3 years) or 4°C (2 years).
  • Safety notes: reported cytotoxic and reproductive toxicity; handle per SDS and use appropriate precautions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.