Learn More
Tisch Scientific Polyvinylidene Fluoride PVDF membrane roll stock, 0.22um, 300mm (width) x 3m (length)
SDP
Catalog No. 502251485
Encompass
Polyvinylidene Fluoride PVDF membrane roll stock, 0.22um, 300mm (width) x 3m (length) with PET substrateThis product has not been established as a suitable replacement for N95 mask material or any other PPE application. In addition, please be aware that we are not able to accept any returned products.
This item is not returnable. View return policy
Catalog No. 50-225-1485 Supplier Tisch Scientific Supplier No. RS20233
Polyvinylidene Fluoride PVDF membrane roll stock, 0.22um, 300mm (width) x 3m (length) with PET substrateThis product has not been established as a suitable replacement for N95 mask material or any other PPE application. In addition, please be aware that we are not able to accept any returned products.