Learn More
Sigma Aldrich Fine Chemicals Biosciences Polyvinylpyrrolidone, 9003-39-8, MFCD00149016, 50 g
Catalog No. NC0277466
Encompass
Linear Formula: (C6H9NO)n, average mol wt 40,000, Synonym: PVP, Polyvidone, Povidone.
Catalog No. NC0277466 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. PVP4050G
Linear Formula: (C6H9NO)n, average mol wt 40,000, Synonym: PVP, Polyvidone, Povidone.