Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Polyvinylpyrrolidone

Catalog No. PVP3605KG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
100 g
500 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PVP3605KG 1 kg
PVP3601KG 500 g
PVP360500G 100 g
PVP360100G 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. PVP3605KG Supplier Sigma Aldrich Supplier No. PVP3605KG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Polyvinyl pyrrolidone [poly(N-vinyl-2-pyrrolidone), PVP] is a stable, bulky, non-toxic, non-ionic polymer widely used in medical, industrial, pharmaceutical, food, cosmetic, and electronic applications. It can be prepared from N-vinyl-2-pyrrolidone by free radical polymerization.PVP can be used as: A stabilizer in the synthesis of metallic nanoparticles (NPs) as it avoids the aggregation of NPs via repulsive forces produced by its hydrophobic carbon chains. A capping agent in the synthesis of rodlike hydroxyapatite (HAp) nanoparticles to control the nucleation and growth of HAp crystal. A synthetic polymer vehicle in the pharmaceutical industry for dispersing and suspending drugs. An emulsifier and disintegrant for solution polymerization.Polyvinylpyrrolidone is a component of Denhardt's Solution and is included at a concentration of 1% (w/v) in the standard 50X stock solution.

Specifications

CAS 9003-39-8
Linear Formula (C6H9NO)n
MDL Number MFCD00149016
Quantity 1 kg
Synonym PVP; Polyvidone; Povidone
Molecular Weight (g/mol) 360000 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.