Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Polyvinylpyrrolidone

Catalog No. 81420500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
81-420-500G 500 g
81-420-100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 81-420-500G Supplier Sigma Aldrich Supplier No. 81420500G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Polyvinylpyrrolidone (PVP), also commercially known as K30, is a water soluble polymer. It has a hygroscopic nature with good adhesive properties. It has a stable pH and has the ability to form transparent films.PVP has a wide range of usage such as:an adhesive for making gluesticks and metal adhesivesa dispersant for ceramicscoatings and inksformation of synthetic fibres and textilesporous membranesPolyvinylpyrrolidone is a component of Denhardt's Solution and is included at a concentration of 1% (w/v) in the standard 50X stock solution.

Specifications

CAS 9003-39-8
Linear Formula (C6H9NO)n
MDL Number MFCD00149016
Quantity 500 g
Synonym Plasdone; PVP; Polyvidone; Povidone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.