Learn More
Medchemexpress LLC Polyvinylpyrrolidone | 9003-39-8 | 25 G
SDP
Catalog No. 5000413916
Encompass_Preferred
Polyvinylpyrrolidone, with an average molecular weight of 360000 Da, is a multifunctional synthetic polymer. It is inert, non-toxic, and biocompatible, making it a versatile excipient widely used to synthesize nanoparticles. It serves in both conventional formulations and novel controlled or targeted delivery systems.
  • Functions as a binder, coating agent, suspending agent, pore-former, solubilizer, and stabilizer.
  • Improves the bioavailability and stability of drugs.
  • Enhances the physicomechanical properties of preparations.
  • Adjusts the release rate of drugs.
  • Prolongs the in vivo circulation time of liposomes.
Catalog No. 50-004-13916 Supplier Medchemexpress LLC Supplier No. 25GHYB1620N
Polyvinylpyrrolidone, with an average molecular weight of 360000 Da, is a multifunctional synthetic polymer. It is inert, non-toxic, and biocompatible, making it a versatile excipient widely used to synthesize nanoparticles. It serves in both conventional formulations and novel controlled or targeted delivery systems.
  • Functions as a binder, coating agent, suspending agent, pore-former, solubilizer, and stabilizer.
  • Improves the bioavailability and stability of drugs.
  • Enhances the physicomechanical properties of preparations.
  • Adjusts the release rate of drugs.
  • Prolongs the in vivo circulation time of liposomes.