Learn More
Medchemexpress LLC Polyvinylpyrrolidone | 9003-39-8 | 25 G
SDP
Catalog No. 5000414267
Encompass_Preferred
Polyvinylpyrrolidone, average Mw~29000 is a multifunctional synthetic polymer with an average molecular weight of 29,000 Da. It serves as a versatile excipient for both conventional formulations and novel controlled or targeted delivery systems, functioning as a binder, coating agent, suspending agent, pore-former, solubilizer, and stabilizer.
  • Serves as a versatile excipient for both conventional formulations and novel controlled or targeted delivery systems.
  • Functions as a binder, coating agent, suspending agent, pore-former, solubilizer, and stabilizer.
  • Shows affinity and adsorption for Indomethacin in vitro.
  • Inhibits drug crystallization and promotes an amorphous state in vitro.
Catalog No. 50-004-14267 Supplier Medchemexpress LLC Supplier No. 25GHYB1620K
Polyvinylpyrrolidone, average Mw~29000 is a multifunctional synthetic polymer with an average molecular weight of 29,000 Da. It serves as a versatile excipient for both conventional formulations and novel controlled or targeted delivery systems, functioning as a binder, coating agent, suspending agent, pore-former, solubilizer, and stabilizer.
  • Serves as a versatile excipient for both conventional formulations and novel controlled or targeted delivery systems.
  • Functions as a binder, coating agent, suspending agent, pore-former, solubilizer, and stabilizer.
  • Shows affinity and adsorption for Indomethacin in vitro.
  • Inhibits drug crystallization and promotes an amorphous state in vitro.