Learn More
Medchemexpress LLC Polyvinylpyrrolidone, average Mw~10000 | 9003-39-8 | 5 G
SDP
Catalog No. 5000412719
Encompass_Preferred
Polyvinylpyrrolidone, average Mw~10000, is a multifunctional synthetic polymer with an average molecular weight of 10000 Da. It is widely used to synthesize nanoparticles and is a versatile excipient for both conventional formulations and novel controlled or targeted delivery systems.
  • Serves as a binder, coating agent, suspending agent, pore-former, solubilizer, and stabilizer.
  • Exhibits rigidity of each monomer and high water solubility.
  • Inhibits the recrystallization of Nimesulide.
  • Enhances the solubility of Curcumin.
Catalog No. 50-004-12719 Supplier Medchemexpress LLC Supplier No. HYB1620J5G
Polyvinylpyrrolidone, average Mw~10000, is a multifunctional synthetic polymer with an average molecular weight of 10000 Da. It is widely used to synthesize nanoparticles and is a versatile excipient for both conventional formulations and novel controlled or targeted delivery systems.
  • Serves as a binder, coating agent, suspending agent, pore-former, solubilizer, and stabilizer.
  • Exhibits rigidity of each monomer and high water solubility.
  • Inhibits the recrystallization of Nimesulide.
  • Enhances the solubility of Curcumin.