Learn More
Medchemexpress LLC Polyvinylpyrrolidone, average Mw~360000 | 9003-39-8 | 100 G
SDP
Catalog No. 5000413286
Encompass_Preferred
Polyvinylpyrrolidone, average Mw~360000 is a multifunctional synthetic polymer with an average molecular weight of 360000 Da. It is widely used to synthesize nanoparticles and is a versatile excipient for both conventional formulations and novel controlled or targeted delivery systems.
  • Serves as a binder, coating agent, suspending agent, pore-former, solubilizer, stabilizer, etc.
  • Useful in pharmacy for improving the bioavailability and stability of drugs.
  • Improves the physicomechanical properties of preparations.
  • Adjusts the release rate of drugs.
  • Prolongs the in vivo circulation time of liposomes.
Catalog No. 50-004-13286 Supplier Medchemexpress LLC Supplier No. HYB1620N100G
Polyvinylpyrrolidone, average Mw~360000 is a multifunctional synthetic polymer with an average molecular weight of 360000 Da. It is widely used to synthesize nanoparticles and is a versatile excipient for both conventional formulations and novel controlled or targeted delivery systems.
  • Serves as a binder, coating agent, suspending agent, pore-former, solubilizer, stabilizer, etc.
  • Useful in pharmacy for improving the bioavailability and stability of drugs.
  • Improves the physicomechanical properties of preparations.
  • Adjusts the release rate of drugs.
  • Prolongs the in vivo circulation time of liposomes.