Learn More
Medchemexpress LLC Polyvinylpyrrolidone, average Mw~40000 | 9003-39-8 | ≥98.0% | 25 G
SDP
Catalog No. 5000414368
Encompass_Preferred
Polyvinylpyrrolidone, average Mw~40000 is a multifunctional synthetic polymer with an average molecular weight of 40,000 Da. It is widely used to synthesize nanoparticles and acts as a versatile excipient for various delivery systems.
  • Functions as a binder, coating agent, suspending agent, pore-former, solubilizer, and stabilizer.
  • Improves drug bioavailability and stability.
  • Enhances physicomechanical properties of preparations.
  • Adjusts the release rate of drugs.
  • Prolongs the in vivo circulation time of liposomes.
Catalog No. 50-004-14368 Supplier Medchemexpress LLC Supplier No. 25GHYB1620L
Polyvinylpyrrolidone, average Mw~40000 is a multifunctional synthetic polymer with an average molecular weight of 40,000 Da. It is widely used to synthesize nanoparticles and acts as a versatile excipient for various delivery systems.
  • Functions as a binder, coating agent, suspending agent, pore-former, solubilizer, and stabilizer.
  • Improves drug bioavailability and stability.
  • Enhances physicomechanical properties of preparations.
  • Adjusts the release rate of drugs.
  • Prolongs the in vivo circulation time of liposomes.