Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polyvinylpyrrolidone-iodine complex

Catalog No. AC229475000
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC229475000 500 g
AC229471000 100 g
AC229470025 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC229475000 Supplier Thermo Scientific Chemicals Supplier No. 229475000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 25655-41-8 | (C6H9NO)nI2 | 364.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25655-41-8
Molecular Formula (C6H9NO)nI2
Molecular Weight (g/mol) 364.95
MDL Number MFCD00084483
InChI Key CPKVUHPKYQGHMW-UHFFFAOYSA-N
Synonym povidone iodine, povidone-iodine, betadine, isodine, pvp iodine, pvp-iodine, pvp-i, isobetadyne, bridine, disphex
PubChem CID 410087
IUPAC Name 1-ethenylpyrrolidin-2-one;molecular iodine
SMILES II.*-CC(-*)N1CCCC1=O

Specifications

Color Brown-Red to Yellow
Loss on Drying 8% max. (5 g, 105°C)
Infrared Spectrum Authentic
Packaging Plastic bottle
Linear Formula (C6H9NO)n·xI
Quantity 500 g
Merck Index 15, 7815
Percent Purity 9.0 to 12.0% (available iodine) (on dried substance)
Physical Form Powder
Chemical Name or Material Polyvinylpyrrolidone-iodine complex
RTECSNumber TR1579600
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.