Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polyvinylpyrrolidone K 30, MP Biomedicals™

Catalog No. ICN19545190 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
ICN19545190 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19545190 Supplier MP Biomedicals, Inc Supplier No. 0219545190
Add to Cart
Add to Cart

Specifications

CAS 9003-39-8
Color White
Density 1.23-1.29g/mL
Molecular Formula (C6H9NO)n
MDL Number MFCD01076626
Quantity 500 g
Synonym n-vinyl-2-pyrrolidone, n-vinylpyrrolidone, 1-vinyl-2-pyrrolidone, 1-vinylpyrrolidin-2-one, n-vinyl-2-pyrrolidinone, vinylpyrrolidone, 2-pyrrolidinone, 1-ethenyl, 1-vinyl-2-pyrrolidinone, n-vinylpyrrolidinone, 1-vinylpyrrolidone
InChI Key WHNWPMSKXPGLAX-UHFFFAOYSA-N
SMILES *-CC(-*)N1CCCC1=O
IUPAC Name 1-ethenylpyrrolidin-2-one
Molecular Weight (g/mol) 111.14
PubChem CID 6917
ChEBI CHEBI:82551
Formula Weight 40000
Physical Form Powder
Chemical Name or Material Polyvinylpyrrolidone K 30
Show More Show Less
Recommended Storage Store at Room Temperature.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.