Learn More
Medchemexpress LLC Polyvinylpyrrolidone K120 | 9003-39-8 | 98.0% | 25 G
SDP
Catalog No. 5000414427
Encompass_Preferred
Polyvinylpyrrolidone K120 is a multifunctional synthetic polymer that exhibits excellent adhesiveness and thickening properties. It serves as a versatile excipient in both conventional formulations and novel controlled or targeted delivery systems. This product can also be utilized in the preparation of drug-loaded hydrogel patches.
  • A non-ionic polymer.
  • Superior properties for various fields including paper, fibers and textiles, ceramics, and pharmaceutics.
  • In pharmacy, its inertness, non-toxicity, and biocompatibility make it a versatile excipient.
  • Can function as a binder, coating agent, suspending agent, pore-former, solubilizer, and stabilizer.
  • Available with different molecular weights and concentrations for various purposes.
Catalog No. 50-004-14427 Supplier Medchemexpress LLC Supplier No. 25GHYB1620I
Polyvinylpyrrolidone K120 is a multifunctional synthetic polymer that exhibits excellent adhesiveness and thickening properties. It serves as a versatile excipient in both conventional formulations and novel controlled or targeted delivery systems. This product can also be utilized in the preparation of drug-loaded hydrogel patches.
  • A non-ionic polymer.
  • Superior properties for various fields including paper, fibers and textiles, ceramics, and pharmaceutics.
  • In pharmacy, its inertness, non-toxicity, and biocompatibility make it a versatile excipient.
  • Can function as a binder, coating agent, suspending agent, pore-former, solubilizer, and stabilizer.
  • Available with different molecular weights and concentrations for various purposes.