Learn More
Medchemexpress LLC Polyvinylpyrrolidone K120 | 9003-39-8 | 98.0% | 50 G
SDP
Catalog No. 5000413126
Encompass_Preferred
Polyvinylpyrrolidone K120 is a multifunctional synthetic polymer that exhibits excellent adhesiveness and thickening properties. It serves as a versatile excipient in both conventional formulations and novel controlled or targeted drug delivery systems, and can be utilized in the preparation of drug-loaded hydrogel patches.
  • Exhibits excellent adhesiveness and thickening properties.
  • Serves as a versatile excipient in conventional formulations and novel drug delivery systems.
  • Can be used to prepare drug-loaded hydrogel patches.
  • Functions as a binder, coating agent, suspending agent, pore-former, solubilizer, and stabilizer.
Catalog No. 50-004-13126 Supplier Medchemexpress LLC Supplier No. HYB1620I50G
Polyvinylpyrrolidone K120 is a multifunctional synthetic polymer that exhibits excellent adhesiveness and thickening properties. It serves as a versatile excipient in both conventional formulations and novel controlled or targeted drug delivery systems, and can be utilized in the preparation of drug-loaded hydrogel patches.
  • Exhibits excellent adhesiveness and thickening properties.
  • Serves as a versatile excipient in conventional formulations and novel drug delivery systems.
  • Can be used to prepare drug-loaded hydrogel patches.
  • Functions as a binder, coating agent, suspending agent, pore-former, solubilizer, and stabilizer.