Learn More
Medchemexpress LLC Polyvinylpyrrolidone K25 | 9003-39-8 | 100MG
SDP
Catalog No. 5000413158
Encompass_Preferred
Polyvinylpyrrolidone K25 is a multifunctional synthetic polymer that serves as a versatile excipient for both conventional formulations and novel controlled or targeted delivery systems. It also functions as a preferable binder for granules and tablets. It prevents or retards drug crystallization from supersaturated systems by inhibiting nucleation and/or crystal growth, which allows for complete drug release.
  • Versatile excipient
  • Binder for granules and tablets
  • Inhibits drug crystallization
  • Allows complete drug release
  • Can be used as a coating agent
  • Functions as a suspending agent
  • Acts as a pore-former
  • Used as a solubilizer
  • Provides stabilization
Catalog No. 50-004-13158 Supplier Medchemexpress LLC Supplier No. HYB1620D100MG
Polyvinylpyrrolidone K25 is a multifunctional synthetic polymer that serves as a versatile excipient for both conventional formulations and novel controlled or targeted delivery systems. It also functions as a preferable binder for granules and tablets. It prevents or retards drug crystallization from supersaturated systems by inhibiting nucleation and/or crystal growth, which allows for complete drug release.
  • Versatile excipient
  • Binder for granules and tablets
  • Inhibits drug crystallization
  • Allows complete drug release
  • Can be used as a coating agent
  • Functions as a suspending agent
  • Acts as a pore-former
  • Used as a solubilizer
  • Provides stabilization