Learn More
Medchemexpress LLC Polyvinylpyrrolidone K25 | 9003-39-8 | 500 MG
SDP
Catalog No. 5000413125
Encompass_Preferred
Polyvinylpyrrolidone K25 is a multifunctional synthetic polymer and a versatile excipient for both conventional formulations and novel controlled or targeted delivery systems. It is also a preferable binder for granules and tablets, preventing or retarding drug crystallization from supersaturated systems by inhibiting nucleation and/or crystal growth, which allows for complete drug release.
  • Multifunctional synthetic polymer
  • Versatile excipient for conventional and novel delivery systems
  • Preferable binder for granules and tablets
  • Prevents or retards drug crystallization
  • Inhibits nucleation and/or crystal growth
  • Enables complete drug release
  • Used in pharmaceutical preparations
  • Functions as a coating, suspending, pore-forming, solubilizing, stabilizing, and inhibiting agent
Catalog No. 50-004-13125 Supplier Medchemexpress LLC Supplier No. HYB1620D500MG
Polyvinylpyrrolidone K25 is a multifunctional synthetic polymer and a versatile excipient for both conventional formulations and novel controlled or targeted delivery systems. It is also a preferable binder for granules and tablets, preventing or retarding drug crystallization from supersaturated systems by inhibiting nucleation and/or crystal growth, which allows for complete drug release.
  • Multifunctional synthetic polymer
  • Versatile excipient for conventional and novel delivery systems
  • Preferable binder for granules and tablets
  • Prevents or retards drug crystallization
  • Inhibits nucleation and/or crystal growth
  • Enables complete drug release
  • Used in pharmaceutical preparations
  • Functions as a coating, suspending, pore-forming, solubilizing, stabilizing, and inhibiting agent