Learn More
Diagnostic Biosystems POLYVUE PLUS MOUSE/RABBIT HRP Mouse/Rabbit PolyVue Plus™ HRP/DAB Detection System
Supplier:  Diagnostic Biosystems PVP100D
Encompass_Preferred
The Mouse/Rabbit PolyVue Plus™ HRP/DAB Detection System is a non-biotin, two-step detection system suitable for labeling antigens in formalin-fixed paraffin-embedded tissues and cryostat sections. The PolyVue Plus™ Detection System may also be used with blood smears, cytosmears, and cell preparations. This system has been developed by directly labeling immunoglobulins with enzymes using a proprietary tandem hyperlabelling technology. This ensures consistent and reproducible immunostaining for all types of nuclear, cytoplasmic and membrane antigens in different types of tissues with significantly lower background than detection systems using biotin and avidin conjugates.
Catalog No. 50-240-7414