Learn More
SPECTRUM QUALITY STANDARDS POLY655/1000-Full Range C5 to C120 (10×1 ml)
Catalog No. NC0875926
Encompass_Preferred
Polywax 655/1000 for D6352 & D7169 which contains a full range of C5 to C120 in a 10×1 ml ampoules set.
Catalog No. NC0875926 Supplier SPECTRUM QUALITY STANDARDS Supplier No. POLY655/1000
Polywax 655/1000 for D6352 & D7169 which contains a full range of C5 to C120 in a 10×1 ml ampoules set.