Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PolyCo™ PolyWear™ Smooth Disposable Gowns

Catalog No. 19125558
Encompass_Preferred
Change view
Click to view available options
Color:
Blue
White
Yellow
Size:
Medium
Large
X-Large
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
19-125-558 Yellow Large
18-568 Blue Large
18-567 Blue Medium
19-125-553 Blue X-Large
19-125-554 White Medium
19-125-555 White Large
19-125-557 Yellow Medium
19-125-559 Yellow X-Large
19-125-556 White X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Catalog No. 19-125-558 Supplier PolyCo™ Supplier No. 10504
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Lightweight, single-layer, low-density polyethylene gown for clinical and laboratory settings.

Lightweight, single-layer, low-density polyethylene gown for clinical and laboratory settings

Heat-sealed seam is behind sleeves—minimizes chance of contamination passing through.

  • Full-length wraparound with open back tailored to eliminate excess material
  • Tear-away collar for quick and easy removal
  • Thumb loop on sleeve keeps cuffs under gloves, enhancing leak protection
  • Gowns are folded for quick shake-out and easy donning
  • Powder blue with smooth matte finish

Order Info

Packed individually or bulk in dispenser box.

Compliance

Complies with FDA regulations. Successfully tested for liquid and aerosol blood resistance under ASTM™ F 1670 Synthetic Blood Penetration Resistance and ASTM 1671 Bacteriophage Penetration Resistance.

Specifications

For Use With (Application) Clinical, Laboratory
Gender Unisex
No. of Pockets None
Size Large
Product Type General Purpose Gown
Material Polyethylene Film (Virgin)
Color Yellow
Disposable Single-use
Sterility Non-sterile
Finish Smooth Matte
Model 10504
Autoclavable Not Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.