Learn More
Apexbio Technology LLC POM 1,50mg CAS# 12141-67-2
SDP
Supplier:  Apexbio Technology LLC B715350
Encompass_Preferred
POM 1, also known as sodium polyoxotungstate and sodium metatungstate, is a novel, potent inhibitor of ecto-nucleoside triphosphate diphosphohydrolases (Ecto-NTPDases). POM 1 exhibits minor selectivity for NTPDase 1 and NTPDase 3 over NTPDase 2, with Ki values being 2.58, 28.8 and 3.26 μM for NTPDase 1 (CD39), NTPDase 2 and NTPDase 3, respectively. Ecto-NTPDases capable of hydrolyzing nucleoside tri- and diphosphates are able to reduce the effective concentration of the released nucleotide, and inhibitors of Ecto-NTPDases can thus serve as valuable tools for amplifying the biological effects triggered by extracellularly released nucleotides.  Other sizes are also available. Please inqury us for quote.
Catalog No. 50-224-0184