Learn More
Selleck Chemical LLC pomaglumetad
SDP
Catalog No. 5000003747
Encompass_Preferred
LY404039 is a potent agonist of recombinant human mGlu2/mGlu3 receptors with Ki of 149 nM/92 nM shows 100-fold selectivity over ionotropic glutamate receptors glutamate transporters and other receptors Phase 3
Catalog No. 50-000-03747 Supplier Selleck Chemical LLC Supplier No. S60015MG
LY404039 is a potent agonist of recombinant human mGlu2/mGlu3 receptors with Ki of 149 nM/92 nM shows 100-fold selectivity over ionotropic glutamate receptors glutamate transporters and other receptors Phase 3