Learn More
Medchemexpress LLC HY-105040C 5mg Medchemexpress, Pomaglumetad methionil (hydrochloride) CAS:635318-26-2 Purity:>98%
SDP
Catalog No. 502080681
Encompass_Preferred
Medchemexpress, HY-105040C 5mg Pomaglumetad methionil (hydrochloride) CAS:635318-26-2 Pomaglumetad methionil hydrochloride (LY2140023 hydrochloride) is an orally active, methionine prodrug of the selective mGlu2/3 receptor agonist LY404039. Pomaglumetad methionil hydrochloride has the potential for schizophrenia research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0681 Supplier Medchemexpress LLC Supplier No. HY105040C5MG
Medchemexpress, HY-105040C 5mg Pomaglumetad methionil (hydrochloride) CAS:635318-26-2 Pomaglumetad methionil hydrochloride (LY2140023 hydrochloride) is an orally active, methionine prodrug of the selective mGlu2/3 receptor agonist LY404039. Pomaglumetad methionil hydrochloride has the potential for schizophrenia research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.