Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pomaglumetad methionil hydrochloride | 635318-26-2 | 98.0% | 402.87 g·mol-1 | C12H19ClN2O7S2 | 100 MG
SDP

Catalog No. 5000243842
Encompass_Preferred

Pomaglumetad methionil hydrochloride is an orally active methionine prodrug of an mGlu2/3 receptor agonist supplied for preclinical neuroscience and pharmacology research. It is provided as a characterized solid with formulation and storage guidance for in vitro and in vivo studies.

  • Purity 98.0%.
  • Solid, off-white to light yellow.
  • Soluble in DMSO (25 mg/mL) with ultrasonic assistance.
  • In vivo formulation protocols provided for aqueous and oil vehicles.
  • Recommended storage 4°C under nitrogen; in solution -80°C for long term.
  • Available in small pack sizes including 100 mg.

Catalog No. 50-002-43842 Supplier Medchemexpress LLC Supplier No. HY105040C100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Pomaglumetad methionil hydrochloride is an orally active methionine prodrug of an mGlu2/3 receptor agonist supplied for preclinical neuroscience and pharmacology research. It is provided as a characterized solid with formulation and storage guidance for in vitro and in vivo studies.

  • Purity 98.0%.
  • Solid, off-white to light yellow.
  • Soluble in DMSO (25 mg/mL) with ultrasonic assistance.
  • In vivo formulation protocols provided for aqueous and oil vehicles.
  • Recommended storage 4°C under nitrogen; in solution -80°C for long term.
  • Available in small pack sizes including 100 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.