Learn More
Medchemexpress LLC Pomaglumetad methionil hydrochloride | 635318-26-2 | 98.0% | 402.87 g/mol | C12H19ClN2O7S2 | 1 ML
SDP
Catalog No. 5000243877
Encompass_Preferred
Pomaglumetad methionil hydrochloride is the methionine prodrug of a selective mGlu2/3 receptor agonist, provided as a 10 mM solution (1 mL) for research applications in pharmacology and neuroscience. It is used to study metabotropic glutamate receptor signaling, behavioral pharmacology, and related biochemical pathways. Handle and store according to the product datasheet and SDS.
  • Hydrochloride salt form.
  • Supplied as a 10 mM solution in 1 mL volume.
  • High purity (98.0%).
  • Molecular weight 402.87 g/mol.
  • Intended for in vitro and in vivo research use only.
  • Refer to safety data sheet for handling and storage.
Catalog No. 50-002-43877 Supplier Medchemexpress LLC Supplier No. HY105040C10MM 1ML
Pomaglumetad methionil hydrochloride is the methionine prodrug of a selective mGlu2/3 receptor agonist, provided as a 10 mM solution (1 mL) for research applications in pharmacology and neuroscience. It is used to study metabotropic glutamate receptor signaling, behavioral pharmacology, and related biochemical pathways. Handle and store according to the product datasheet and SDS.
  • Hydrochloride salt form.
  • Supplied as a 10 mM solution in 1 mL volume.
  • High purity (98.0%).
  • Molecular weight 402.87 g/mol.
  • Intended for in vitro and in vivo research use only.
  • Refer to safety data sheet for handling and storage.