Learn More
TENOVA PHARMACEUTICALS INC Pomalidomide-PEG1-C2-amine HCl, 100 mg
Catalog No. NC2861779
Encompass
Degrader building block for targeted protein degradation. Contains an E3 ligase ligand with a linker and a function group, which can be conjugated to target protein ligands through straightforward chemical reactions.
Catalog No. NC2861779 Supplier TENOVA PHARMACEUTICALS INC Supplier No. T52006100MG
Degrader building block for targeted protein degradation. Contains an E3 ligase ligand with a linker and a function group, which can be conjugated to target protein ligands through straightforward chemical reactions.