Learn More
Medchemexpress LLC Pomalidomi 10 mM * 1 mL in DMSO Solution
SDP
Catalog No. NC3451842
Encompass_Preferred
Pomalidomide, the third-generation immunomodulatory agent, acts as molecular glue. Pomalidomide interacts with the E3 ligase cereblon and induces degradation of essential Ikaros transcription factors.
Catalog No. NC3451842 Supplier Medchemexpress LLC Supplier No. HY1098410MM1MLDMSO
Pomalidomide, the third-generation immunomodulatory agent, acts as molecular glue. Pomalidomide interacts with the E3 ligase cereblon and induces degradation of essential Ikaros transcription factors.