Learn More
Cayman Chemical PomalIdomID 4alkylC6amIn 100mg
SDP
Catalog No. 502942059
Encompass_Preferred
A PROTAC building block; has been used in the synthesis of PROTACs that drive HDAC8 degradation
Catalog No. 50-294-2059 Supplier Cayman Chemical Supplier No. 39985100 MG
A PROTAC building block; has been used in the synthesis of PROTACs that drive HDAC8 degradation