Learn More
Medchemexpress LLC HY-10984 200mg , Pomalidomide CC-4047 CAS:19171-19-8 Purity:98%
SDP
Catalog No. 501492551
Encompass_Preferred
Medchemexpress, HY-10984 200mg Pomalidomide CC-4047 CAS:19171-19-8 Formula:C13H11N3O4 IC50: 13 nM (TNF-α, in PBMCs) Purity:98% Pomalidomide is a known inhibitor of TNF-α release in LPS stimulated human PBMC with IC 50 of 13 nM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2551 Supplier Medchemexpress LLC Supplier No. HY10984200MG
Medchemexpress, HY-10984 200mg Pomalidomide CC-4047 CAS:19171-19-8 Formula:C13H11N3O4 IC50: 13 nM (TNF-α, in PBMCs) Purity:98% Pomalidomide is a known inhibitor of TNF-α release in LPS stimulated human PBMC with IC 50 of 13 nM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.