Learn More
Medchemexpress LLC Pomalidomide 4'-alkylC5-acid | 2225940-49-6 | 98.8% | 250 MG
SDP
Catalog No. 5000342807
Encompass_Preferred
Pomalidomide 4'-alkylC5-acid is an E3 ligase ligand-linker conjugate. It contains a cereblon (CRBN) ligand, which is Pomalidomide based, and a linker. This compound can be utilized in the synthesis of PROTAC (Proteolysis Targeting Chimeras). PROTACs are composed of two distinct ligands connected by a linker, where one ligand targets an E3 ubiquitin ligase and the other targets the protein of interest. These molecules function by harnessing the intracellular ubiquitin-proteasome system to achieve the selective degradation of target proteins.
  • Can be utilized in the synthesis of PROTAC
  • Contains a cereblon (CRBN) ligand
  • Contains a linker
  • Targets an E3 ubiquitin ligase
  • Targets the protein of interest
  • Harnesses the intracellular ubiquitin-proteasome system
  • Achieves selective degradation of target proteins
Catalog No. 50-003-42807 Supplier Medchemexpress LLC Supplier No. 250MGHY130737
Pomalidomide 4'-alkylC5-acid is an E3 ligase ligand-linker conjugate. It contains a cereblon (CRBN) ligand, which is Pomalidomide based, and a linker. This compound can be utilized in the synthesis of PROTAC (Proteolysis Targeting Chimeras). PROTACs are composed of two distinct ligands connected by a linker, where one ligand targets an E3 ubiquitin ligase and the other targets the protein of interest. These molecules function by harnessing the intracellular ubiquitin-proteasome system to achieve the selective degradation of target proteins.
  • Can be utilized in the synthesis of PROTAC
  • Contains a cereblon (CRBN) ligand
  • Contains a linker
  • Targets an E3 ubiquitin ligase
  • Targets the protein of interest
  • Harnesses the intracellular ubiquitin-proteasome system
  • Achieves selective degradation of target proteins