Learn More
Medchemexpress LLC Pomalidomide 4'-alkylC5-acid | 2225940-49-6 | 98.8% | 50 MG
SDP
Catalog No. 5000339898
Encompass_Preferred
Pomalidomide 4'-alkylC5-acid is an E3 ligase ligand-linker conjugate. It contains a Pomalidomide-based cereblon (CRBN) ligand and a linker. This product can be utilized in the synthesis of PROTAC.
  • Serves as an E3 ligase ligand-linker conjugate for the synthesis of PROTACs.
  • Utilizes the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
Catalog No. 50-003-39898 Supplier Medchemexpress LLC Supplier No. HY13073750MG
Pomalidomide 4'-alkylC5-acid is an E3 ligase ligand-linker conjugate. It contains a Pomalidomide-based cereblon (CRBN) ligand and a linker. This product can be utilized in the synthesis of PROTAC.
  • Serves as an E3 ligase ligand-linker conjugate for the synthesis of PROTACs.
  • Utilizes the intracellular ubiquitin-proteasome system to selectively degrade target proteins.