Learn More
Medchemexpress LLC Pomalidomide 4'-PEG3-azide | 2271036-46-3 | 96.3% | 250 MG
SDP
Catalog No. 5000343006
Encompass_Preferred
Pomalidomide 4'-PEG3-azide is a synthesized E3 ligase ligand-linker conjugate that incorporates the Pomalidomide-based cereblon ligand and a linker. It can be used for the synthesis of iRucaparib-TP3 (Compound 3), a highly efficient PARP1 degrader based on Rucaparib using the PROTAC approach.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates a Pomalidomide-based cereblon ligand and a linker
  • Can be used for the synthesis of efficient PARP1 degraders
  • Click chemistry reagent containing an azide group
  • Capable of copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with alkyne groups
  • Capable of strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups
Catalog No. 50-003-43006 Supplier Medchemexpress LLC Supplier No. 250MGHY130652
Pomalidomide 4'-PEG3-azide is a synthesized E3 ligase ligand-linker conjugate that incorporates the Pomalidomide-based cereblon ligand and a linker. It can be used for the synthesis of iRucaparib-TP3 (Compound 3), a highly efficient PARP1 degrader based on Rucaparib using the PROTAC approach.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates a Pomalidomide-based cereblon ligand and a linker
  • Can be used for the synthesis of efficient PARP1 degraders
  • Click chemistry reagent containing an azide group
  • Capable of copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with alkyne groups
  • Capable of strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups