Learn More
Medchemexpress LLC HY-131647 50mg Medchemexpress, Pomalidomide 4'-PEG5-acid CAS:2139348-63-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13164750MG
Encompass_Preferred
Medchemexpress, HY-131647 50mg Pomalidomide 4'-PEG5-acid CAS:2139348-63-1 Pomalidomide 4'-PEG5-acid (Pomalidomide-PEG5-CO2H) is a synthesized E3 ligase ligand-linker conjugate that incorporates the Pomalidomide based cereblon ligand and 5-unit PEG linker used in PROTAC technology. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1292