Learn More
Cayman Chemical PomalIdomIde 4-PEG5-amInh 50mg
SDP
Catalog No. 502942175
Encompass_Preferred
A PEGylated form of pomalidomide; has been conjugated to various ligands in the development of PROTACs
Catalog No. 50-294-2175 Supplier Cayman Chemical Supplier No. 3998750 MG
A PEGylated form of pomalidomide; has been conjugated to various ligands in the development of PROTACs