Learn More
Cayman Chemical PomalIdomIde 4alkylC6amIn 25mg
SDP
Catalog No. 502942447
Encompass_Preferred
A PROTAC building block; has been used in the synthesis of PROTACs that drive HDAC8 degradation
Catalog No. 50-294-2447 Supplier Cayman Chemical Supplier No. 3998525 MG
A PROTAC building block; has been used in the synthesis of PROTACs that drive HDAC8 degradation