Learn More
Medchemexpress LLC Pomalidomide-5-C10-NH2 hydrochloride | 2863635-00-9 | 95.0% | 464.99 | C23H33ClN4O4 | 10 MG
SDP
Catalog No. 5000287469
Encompass_Preferred
Pomalidomide-5-C10-NH2 hydrochloride is a pomalidomide-derived cereblon (CRBN) ligand supplied as the hydrochloride salt for use in targeted protein degradation research. It is intended as a building block for PROTAC synthesis and related biochemical studies, provided in small research quantities with defined purity and molecular properties.
  • Acts as a cereblon E3 ligase recruiter for PROTAC design.
  • Provided as the hydrochloride salt for improved stability and handling.
  • Offered in small-milligram quantities suitable for medicinal chemistry work.
  • Characterized by specified purity and molecular weight for reproducible results.
  • Compatible with standard organic solvents used in synthesis and assays.
Catalog No. 50-002-87469 Supplier Medchemexpress LLC Supplier No. HY15526610MG
Pomalidomide-5-C10-NH2 hydrochloride is a pomalidomide-derived cereblon (CRBN) ligand supplied as the hydrochloride salt for use in targeted protein degradation research. It is intended as a building block for PROTAC synthesis and related biochemical studies, provided in small research quantities with defined purity and molecular properties.
  • Acts as a cereblon E3 ligase recruiter for PROTAC design.
  • Provided as the hydrochloride salt for improved stability and handling.
  • Offered in small-milligram quantities suitable for medicinal chemistry work.
  • Characterized by specified purity and molecular weight for reproducible results.
  • Compatible with standard organic solvents used in synthesis and assays.