Learn More
Medchemexpress LLC Pomalidomide-5-C11-NH2 hydrochloride | 98.2% | 479.01 g/mol | C24H35ClN4O4 | 10 MG
SDP
Catalog No. 5000286878
Encompass_Preferred
Pomalidomide-5-C11-NH2 hydrochloride is a pomalidomide-based cereblon (CRBN) ligand used to recruit the CRBN E3 ligase in targeted protein degradation studies and other chemical biology applications. Supplied as the hydrochloride salt, the compound is provided as a light blue to blue solid with high reported purity and is packaged in small milligram quantities for research use. Storage and handling instructions are provided to preserve stability.
  • High purity (98.2%) suitable for research-grade applications.
  • Hydrochloride salt form for improved stability and handling.
  • Light blue to blue solid for straightforward visual identification.
  • Available in small milligram packaging for screening and synthesis workflows.
  • Storage guidance provided to maintain compound integrity.
Catalog No. 50-002-86878 Supplier Medchemexpress LLC Supplier No. HY15526710MG
Pomalidomide-5-C11-NH2 hydrochloride is a pomalidomide-based cereblon (CRBN) ligand used to recruit the CRBN E3 ligase in targeted protein degradation studies and other chemical biology applications. Supplied as the hydrochloride salt, the compound is provided as a light blue to blue solid with high reported purity and is packaged in small milligram quantities for research use. Storage and handling instructions are provided to preserve stability.
  • High purity (98.2%) suitable for research-grade applications.
  • Hydrochloride salt form for improved stability and handling.
  • Light blue to blue solid for straightforward visual identification.
  • Available in small milligram packaging for screening and synthesis workflows.
  • Storage guidance provided to maintain compound integrity.