Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pomalidomide-5'-PEG5-C2-COOH | 2412056-50-7 | 565.57 | 100 MG
SDP

Catalog No. 5000362212
Encompass_Preferred

Pomalidomide-5'-PEG5-C2-COOH is an active compound and drug derivative used for various biochemical research purposes. This compound requires specific storage conditions to maintain its stability.

  • Active compound and drug derivative
  • Store at -20°C, protected from light, and under nitrogen
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light and under nitrogen
  • Soluble in DMSO at 100 mg/mL (176.81 mM; requires ultrasonic)
  • Molecular formula: C₂₆H₃₅N₃O₁₁

Catalog No. 50-003-62212 Supplier Medchemexpress LLC Supplier No. HY148484100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pomalidomide-5'-PEG5-C2-COOH is an active compound and drug derivative used for various biochemical research purposes. This compound requires specific storage conditions to maintain its stability.

  • Active compound and drug derivative
  • Store at -20°C, protected from light, and under nitrogen
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light and under nitrogen
  • Soluble in DMSO at 100 mg/mL (176.81 mM; requires ultrasonic)
  • Molecular formula: C₂₆H₃₅N₃O₁₁

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.