Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pomalidomide-5'-PEG5-C2-COOH | 2412056-50-7 | 98.1% | 565.57 | 10 MM * 1 ML
SDP

Catalog No. 5000362211
Encompass_Preferred

Pomalidomide-5'-PEG5-C2-COOH is an active compound for the research of various biochemicals. This product is for research purposes only.

  • Purity of 98.1%
  • CAS number: 2412056-50-7
  • Molecular weight: 565.57
  • Recommended storage at -20°C, protected from light, and under nitrogen
  • Recommended storage in solvent at -80°C for 6 months or -20°C for 1 month, protected from light, and under nitrogen
  • Solubility in DMSO: 100 mg/mL

Catalog No. 50-003-62211 Supplier Medchemexpress LLC Supplier No. HY14848410MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pomalidomide-5'-PEG5-C2-COOH is an active compound for the research of various biochemicals. This product is for research purposes only.

  • Purity of 98.1%
  • CAS number: 2412056-50-7
  • Molecular weight: 565.57
  • Recommended storage at -20°C, protected from light, and under nitrogen
  • Recommended storage in solvent at -80°C for 6 months or -20°C for 1 month, protected from light, and under nitrogen
  • Solubility in DMSO: 100 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.