Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pomalidomide-5'-PEG5-C2-COOH | 2412056-50-7 | 98.1% | 565.57 | 25 MG
SDP

Catalog No. 5000363506
Encompass_Preferred

Pomalidomide-5'-PEG5-C2-COOH is an active compound used for the research of various biochemicals. It is designed for research purposes only.

  • Active compound for biochemical studies
  • Targeted as a drug derivative
  • Appears as an oil, green to dark green in color
  • Molecular weight: 565.57
  • Purity: 98.1%
  • Store at -20°C, protected from light and under nitrogen

Catalog No. 50-003-63506 Supplier Medchemexpress LLC Supplier No. 25MGHY148484
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pomalidomide-5'-PEG5-C2-COOH is an active compound used for the research of various biochemicals. It is designed for research purposes only.

  • Active compound for biochemical studies
  • Targeted as a drug derivative
  • Appears as an oil, green to dark green in color
  • Molecular weight: 565.57
  • Purity: 98.1%
  • Store at -20°C, protected from light and under nitrogen

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.