Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pomalidomide-5'-peg8-c2-cooh | 2138440-78-3 | 98.2% | 697.73 | C32H47N3O14 | 10 MG
SDP

Catalog No. 5000204007
Encompass_Preferred

Pomalidomide-5'-PEG8-C2-COOH is an E3 ligase ligand-linker conjugate that links a pomalidomide E3 ligase ligand to an eight-unit polyethylene glycol (PEG8) spacer terminated with a two-carbon carboxylic acid. It is intended as a building block for PROTAC synthesis and other E3 ligase recruitment applications.

  • Used as a building block for PROTAC synthesis and E3 ligase recruitment studies.
  • Conjugate of pomalidomide and a PEG8 linker with a C2 carboxylic acid terminus.
  • High purity: 98.2%.
  • Molecular weight 697.73 g/mol; formula C32H47N3O14.
  • Soluble in DMSO (100 mg/mL) and suitable in vivo formulations (≥2.5 mg/mL).
  • Storage: pure form -20°C (3 years) or 4°C (2 years).

Catalog No. 50-002-04007 Supplier Medchemexpress LLC Supplier No. HYW58002210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pomalidomide-5'-PEG8-C2-COOH is an E3 ligase ligand-linker conjugate that links a pomalidomide E3 ligase ligand to an eight-unit polyethylene glycol (PEG8) spacer terminated with a two-carbon carboxylic acid. It is intended as a building block for PROTAC synthesis and other E3 ligase recruitment applications.

  • Used as a building block for PROTAC synthesis and E3 ligase recruitment studies.
  • Conjugate of pomalidomide and a PEG8 linker with a C2 carboxylic acid terminus.
  • High purity: 98.2%.
  • Molecular weight 697.73 g/mol; formula C32H47N3O14.
  • Soluble in DMSO (100 mg/mL) and suitable in vivo formulations (≥2.5 mg/mL).
  • Storage: pure form -20°C (3 years) or 4°C (2 years).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.