Learn More
Medchemexpress LLC Pomalidomide-5'-PEG8-C2-COOH | 2138440-78-3 | 98.2% | 697.73 | C32H47N3O14 | 50 MG
SDP
Catalog No. 5000204002
Encompass_Preferred
Pomalidomide-5'-PEG8-C2-COOH is an E3 ligase ligand-linker conjugate intended as a building block for proteolysis-targeting chimera (PROTAC) synthesis. The molecule combines a pomalidomide-derived warhead with an eight-unit polyethylene glycol (PEG8) spacer and a terminal two-carbon carboxylic acid for coupling, supplied as a high-purity solid for research use.
  • Designed for PROTAC construction and E3 ligase engagement.
  • Pomalidomide-derived warhead with PEG8 spacer and terminal carboxylic acid for conjugation.
  • High reported purity suitable for synthetic applications.
  • Soluble in DMSO at high concentration for stock solutions.
  • Recommended long-term storage at low temperatures to preserve integrity.
Catalog No. 50-002-04002 Supplier Medchemexpress LLC Supplier No. HYW58002250MG
Pomalidomide-5'-PEG8-C2-COOH is an E3 ligase ligand-linker conjugate intended as a building block for proteolysis-targeting chimera (PROTAC) synthesis. The molecule combines a pomalidomide-derived warhead with an eight-unit polyethylene glycol (PEG8) spacer and a terminal two-carbon carboxylic acid for coupling, supplied as a high-purity solid for research use.
  • Designed for PROTAC construction and E3 ligase engagement.
  • Pomalidomide-derived warhead with PEG8 spacer and terminal carboxylic acid for conjugation.
  • High reported purity suitable for synthetic applications.
  • Soluble in DMSO at high concentration for stock solutions.
  • Recommended long-term storage at low temperatures to preserve integrity.