Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pomalidomide-5'-PEG8-C2-COOH | 2138440-78-3 | 98.2% | 697.73 g/mol | C32H47N3O14 | 25 MG
SDP

Catalog No. 5000203876
Encompass_Preferred

Pomalidomide-5'-PEG8-C2-COOH is an E3 ligase ligand-linker conjugate designed as a building block for PROTAC synthesis. It combines a pomalidomide CRBN-binding moiety with an 8-unit PEG linker and a C2 carboxylic acid handle to enable conjugation to target ligands.

  • useful building block for proteolysis-targeting chimeras (PROTACs).
  • high purity suitable for synthetic applications.
  • soluble in DMSO at approximately 100 mg/mL.
  • molecular weight about 697.73 g/mol and formula C32H47N3O14.
  • storage: pure form -20°C (3 years) or 4°C (2 years).

Catalog No. 50-002-03876 Supplier Medchemexpress LLC Supplier No. HYW58002225MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pomalidomide-5'-PEG8-C2-COOH is an E3 ligase ligand-linker conjugate designed as a building block for PROTAC synthesis. It combines a pomalidomide CRBN-binding moiety with an 8-unit PEG linker and a C2 carboxylic acid handle to enable conjugation to target ligands.

  • useful building block for proteolysis-targeting chimeras (PROTACs).
  • high purity suitable for synthetic applications.
  • soluble in DMSO at approximately 100 mg/mL.
  • molecular weight about 697.73 g/mol and formula C32H47N3O14.
  • storage: pure form -20°C (3 years) or 4°C (2 years).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.