Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pomalidomide 5'-piperazine-4-methylpiperidine (dihydrochloride) | 2758431-93-3 | 98.1% | 512.43 | 250 MG
SDP

Catalog No. 5000392310
Encompass_Preferred

Pomalidomide 5'-piperazine-4-methylpiperidine dihydrochloride is a chemical offered by MedChemExpress for laboratory research. This product is stable under recommended storage conditions and is intended for use by qualified scientists in appropriately equipped facilities.

  • For laboratory chemical research
  • Stable under recommended storage conditions
  • Light yellow to brown solid
  • Purity of 98.08%
  • Recommended storage at 4°C, sealed, away from moisture

Catalog No. 50-003-92310 Supplier Medchemexpress LLC Supplier No. 250MGHY160244
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pomalidomide 5'-piperazine-4-methylpiperidine dihydrochloride is a chemical offered by MedChemExpress for laboratory research. This product is stable under recommended storage conditions and is intended for use by qualified scientists in appropriately equipped facilities.

  • For laboratory chemical research
  • Stable under recommended storage conditions
  • Light yellow to brown solid
  • Purity of 98.08%
  • Recommended storage at 4°C, sealed, away from moisture

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.