Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pomalidomide 5'-piperazine-4-methylpiperidine (dihydrochloride) | 2758431-93-3 | 98.1% | 512.43 | 5 MG
SDP

Catalog No. 5000392298
Encompass_Preferred

Pomalidomide 5'-piperazine-4-methylpiperidine (dihydrochloride) is a chemical compound intended for research use. This compound is stable under recommended storage conditions and has a molecular weight of 512.43 with a purity of 98.08%. It is suitable for laboratory chemical applications and the manufacture of substances.

  • For research use only
  • Stable under recommended storage conditions
  • Store at 4°C, sealed, away from moisture
  • Suitable for laboratory chemicals and manufacture of substances
  • Molecular weight of 512.43
  • Purity of 98.08%

Catalog No. 50-003-92298 Supplier Medchemexpress LLC Supplier No. HY1602445MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pomalidomide 5'-piperazine-4-methylpiperidine (dihydrochloride) is a chemical compound intended for research use. This compound is stable under recommended storage conditions and has a molecular weight of 512.43 with a purity of 98.08%. It is suitable for laboratory chemical applications and the manufacture of substances.

  • For research use only
  • Stable under recommended storage conditions
  • Store at 4°C, sealed, away from moisture
  • Suitable for laboratory chemicals and manufacture of substances
  • Molecular weight of 512.43
  • Purity of 98.08%

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.